Learn More
Electron Microscopy Sciences High Density Polyethylene Scintilation Vials
SDP
Supplier:  Electron Microscopy Sciences 72632PE
Encompass_Preferred
Opaque, high-density polyethylene vials are 20ml in capacity and they measure 27mm diameter x 61mm height (with cap). The screw caps are 22-400 and they are made from polypropylene with a foam- polyethylene liner.
Catalog No. 50-365-858