Learn More
Analytical Sales & Services Scintillation Vial Kit: 20mL Clear Glass Screw Vial with Solid Urea Polyethylene Cone Lined Cap
Catalog No. 5000805428
Encompass
Advantage™ Assembled Scintillation Vial Kit\n\nContains: 20mL (22-400) Clear Borosilicate Glass Screw Vial with Solid Urea Polyethylene Cone Lined Cap
Catalog No. 50-008-05428 Supplier Analytical Sales & Services Supplier No. ADV33320
Advantage™ Assembled Scintillation Vial Kit\n\nContains: 20mL (22-400) Clear Borosilicate Glass Screw Vial with Solid Urea Polyethylene Cone Lined Cap