Learn More
Research Products International Corp Scintillation Vial Rack, 50 Capacity, 5/cs
SDP
Catalog No. 501977444
Encompass
Racks are manufactured from polypropylene and can be easily cleaned in an automatic washer or autoclaved. The racks are sturdy and can be stacked, even when the vials are in place. Each well has an opening in the bottom to facilitate drainage. Autoclavable at 121°.
Catalog No. 50-197-7444 Supplier Research Products International Corp Supplier No. 148806
Racks are manufactured from polypropylene and can be easily cleaned in an automatic washer or autoclaved. The racks are sturdy and can be stacked, even when the vials are in place. Each well has an opening in the bottom to facilitate drainage. Autoclavable at 121°.