Learn More
Research Products International Corp Acrylic Storage Container with Cardboard Tray and Partition Divider for 20ml Vials
SDP
Catalog No. 50212685
Encompass
Versatile acrylic container provides a convenient system for storing or transporting scintillation vials. Container holds one tray of scintillation vials (100 x 20 ml capacity) and can be stacked for maximum storage space. Removable end panel allows vial tray to be easily inserted and removed. Constructed of clear acrylic that also provides beta radiation shielding.
Supplied with one cardboard tray and partition.
Catalog No. 50-212-685 Supplier Research Products International Corp Supplier No. 347000
Versatile acrylic container provides a convenient system for storing or transporting scintillation vials. Container holds one tray of scintillation vials (100 x 20 ml capacity) and can be stacked for maximum storage space. Removable end panel allows vial tray to be easily inserted and removed. Constructed of clear acrylic that also provides beta radiation shielding.
Supplied with one cardboard tray and partition.