Learn More
Research Products International Corp Scintillation Vial Trays with Partitions for Mini-Vials, 200 Vial Capacity, 5 Sets per Package
SDP
Catalog No. 50998594
Encompass
Mini Scintillation Vial Trays are ideal for organizing, storing and transporting 5-8ml sample minivials. Cardboard tray includes 10 x 20 array partition for conveniently holding up to 200 minivials.
Supplied as 5 sets of trays and partitions per package
Catalog No. 50-998-594 Supplier Research Products International Corp Supplier No. 347005
Mini Scintillation Vial Trays are ideal for organizing, storing and transporting 5-8ml sample minivials. Cardboard tray includes 10 x 20 array partition for conveniently holding up to 200 minivials.
Supplied as 5 sets of trays and partitions per package