Learn More
Sigma Aldrich Fine Chemicals Biosciences Scintillation vials genera
Catalog No. 502906024
Encompass
General purpose vials can be used for liquid scintillation counting gamma counting chromatography sample storage and culturing. fits LKB and Packard Varisette counters. Vials are securely sealed by snapping the cap on. Cap removal requires a simple 1/4 twist reducing the chance of spillage associated with pull of caps. Polyethylene vials are supplied with polypropylene caps. Polypropylene vials are supplied with polyethylene caps.
Catalog No. 50-290-6024 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z3768171PAK
General purpose vials can be used for liquid scintillation counting gamma counting chromatography sample storage and culturing. fits LKB and Packard Varisette counters. Vials are securely sealed by snapping the cap on. Cap removal requires a simple 1/4 twist reducing the chance of spillage associated with pull of caps. Polyethylene vials are supplied with polypropylene caps. Polypropylene vials are supplied with polyethylene caps.