Learn More
Sigma Aldrich Fine Chemicals Biosciences Polypropylene Scintillation Vials | Standard Size | 1 Package | Quantity: 100 PCS
Catalog No. NC2335435
Encompass
"Scintillation vials polypropylene | Capacity: 6.5mL | Dimensions: 28mm x 57.5mm | Compatible with: LKB, Packard | Reusable | Standard size | Suitable for: liquid chromatography, sample storage | Qty: 1 pack"
Catalog No. NC2335435 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z3768251PAK
"Scintillation vials polypropylene | Capacity: 6.5mL | Dimensions: 28mm x 57.5mm | Compatible with: LKB, Packard | Reusable | Standard size | Suitable for: liquid chromatography, sample storage | Qty: 1 pack"