Learn More
Research Products International Corp Wide-Mouth Copolymer Plastic Vials, 100 per Tray, 500 per Case
SDP
Catalog No. 501367486
Encompass
Wide-mouth opening (23mm) for large samples such as filter discs. Constructed of translucent copolymer polypropylene with exceptional clarity. Supplied with a white polyethylene cap that seals with a 1/4 turn. Packaged in 100 cell partition trays. Capacity: 20 ml; Cap Size: 26mm; Dimensions (Cap On): 26x61mm.
Catalog No. 50-136-7486 Supplier Research Products International Corp Supplier No. 121039TP
Wide-mouth opening (23mm) for large samples such as filter discs. Constructed of translucent copolymer polypropylene with exceptional clarity. Supplied with a white polyethylene cap that seals with a 1/4 turn. Packaged in 100 cell partition trays. Capacity: 20 ml; Cap Size: 26mm; Dimensions (Cap On): 26x61mm.