Learn More
Electron Microscopy Sciences SEM Scintillator YAP 19.8x1 mm
SDP
Catalog No. 501900130
Encompass_Preferred
YAP (Yttrium Aluminum Perovskite activated by Ce3+) shares the same advantages as YAG but is more efficient in terms of light output than YAG crystals. As well, the emission spectrum peaks at about 378nm, and this corresponds closely to the maximum sensitivity of the S11 photocathode which in general is used in most scanning electron microscopes. (There would be more improvement in signal by using a YAP crystal rather than YAG in these microscopes). The decay time of YAP (40ns) is faster than YAG (80ns) so its overall performance is superior to YAG.
YAP crystals should be coated with 50nm of aluminum prior to use. Note: P-47 is usually in-stock. YAG and YAP delivery time is 6-8 weeks. Use with Hitachi w/metal ring.
Catalog No. 50-190-0130 Supplier Electron Microscopy Sciences Supplier No. 82089
YAP (Yttrium Aluminum Perovskite activated by Ce3+) shares the same advantages as YAG but is more efficient in terms of light output than YAG crystals. As well, the emission spectrum peaks at about 378nm, and this corresponds closely to the maximum sensitivity of the S11 photocathode which in general is used in most scanning electron microscopes. (There would be more improvement in signal by using a YAP crystal rather than YAG in these microscopes). The decay time of YAP (40ns) is faster than YAG (80ns) so its overall performance is superior to YAG.
YAP crystals should be coated with 50nm of aluminum prior to use. Note: P-47 is usually in-stock. YAG and YAP delivery time is 6-8 weeks. Use with Hitachi w/metal ring.