Learn More
Electron Microscopy Sciences SEM Scintillator YAP 20.0x2 mm
SDP
Catalog No. 501900132
Encompass_Preferred
YAP (Yttrium Aluminum Perovskite activated by Ce3+) shares the same advantages as YAG but is more efficient in terms of light output than YAG crystals. As well, the emission spectrum peaks at about 378nm, and this corresponds closely to the maximum sensitivity of the S11 photocathode which in general is used in most scanning electron microscopes. (There would be more improvement in signal by using a YAP crystal rather than YAG in these microscopes). The decay time of YAP (40ns) is faster than YAG (80ns) so its overall performance is superior to YAG.
YAP crystals should be coated with 50nm of aluminum prior to use. Note: P-47 is usually in-stock. YAG and YAP delivery time is 6-8 weeks. Use with Philips scanning electron microscopes.
Catalog No. 50-190-0132 Supplier Electron Microscopy Sciences Supplier No. 82092
Add to Cart
Add to Cart
YAP (Yttrium Aluminum Perovskite activated by Ce3+) shares the same advantages as YAG but is more efficient in terms of light output than YAG crystals. As well, the emission spectrum peaks at about 378nm, and this corresponds closely to the maximum sensitivity of the S11 photocathode which in general is used in most scanning electron microscopes. (There would be more improvement in signal by using a YAP crystal rather than YAG in these microscopes). The decay time of YAP (40ns) is faster than YAG (80ns) so its overall performance is superior to YAG.
YAP crystals should be coated with 50nm of aluminum prior to use. Note: P-47 is usually in-stock. YAG and YAP delivery time is 6-8 weeks. Use with Philips scanning electron microscopes.