Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ScintiSafe™ Econo F Cocktail (Scintanalyzed™), Fisher Chemical

Catalog No. SX225 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
5 L
Packaging:
Black Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SX22-5 5 L Black Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SX22-5

missing translation for 'validMainframeMsg'

Specifications

Boiling Point 280°C
Chemical Name or Material ScintiSafe™ Econo F Cocktail
Melting Point -70°C
Physical Form Liquid
Density 0.86g/cm³
Color Colorless
Quantity 5 L
MDL Number MFCD00242091
Packaging Black Poly Bottle
Synonym LAB-based (Linear Alkylbenzene)
Grade Scintanalyzed
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.