Learn More
Electron Microscopy Sciences Wheaton 180 Glass Scintilation Vials
SDP
Supplier:  Electron Microscopy Sciences 72634
Encompass_Preferred
Wheaton "180" low potassium borosilicate glass meets ASTM Type I Class A and USP Type I standards. Urea screw caps with Poly-seal cone liner. Measures 28mm diameter x 57mm height (cap off), and 61mm height (cap on), size 20.
Catalog No. 50-189-9972