Learn More
Electron Microscopy Sciences Polyethylene Terephthalate Scintilation Vials
SDP
Catalog No. 501899974
Encompass_Preferred
Made from polypropylene with a foamed polyethylene liner. Lightweight, unbreakable, with the clarity of glass. Low permeability to solvents and minimal background count. Measures 27mm diameter, 61mm height, caps are 22-400.
Catalog No. 50-189-9974 Supplier Electron Microscopy Sciences Supplier No. 72634PT
Made from polypropylene with a foamed polyethylene liner. Lightweight, unbreakable, with the clarity of glass. Low permeability to solvents and minimal background count. Measures 27mm diameter, 61mm height, caps are 22-400.