Learn More
PARKER HANNIFIN CORP SCIPRES PRSSR SNSR 1/2 BRB 5PK
SDP
Catalog No. NC0087853
Encompass
1/2" Barb for tube size range ID 0.50, 17 LPM, 60 PSI. SciLog� SciPres� scalable pressure sensors are designed for incorporation into single-use purification platforms, available in five sizes. Pre-calibrated and gamma stable, calibration and identity information is stored on the sensor's chip.
Catalog No. NC0087853 Supplier PARKER HANNIFIN CORP Supplier No. 080695PSX5
1/2" Barb for tube size range ID 0.50, 17 LPM, 60 PSI. SciLog� SciPres� scalable pressure sensors are designed for incorporation into single-use purification platforms, available in five sizes. Pre-calibrated and gamma stable, calibration and identity information is stored on the sensor's chip.