Learn More
PARKER HANNIFIN CORP SCIPRES PRSSR SNSR 3/8 BRB 5PK
SDP
Catalog No. NC0087851
Encompass
The Scilog� Scipres� Combines Pressure Sensing Capabilities And The Convenience Of Disposability With Easy Setup. Each Sensor Is Preprogrammed With A Unique Id For Easy Traceability And Data Documentation When Combined With The Scilog� Scidoc Software. Factory Calibration Data Is Also Stored On Each Sensor'S Chip For Out-Of-Box, Plug And Play Use. Connection Type: 3/8" Barb Tube Size Range: Id 0.31" To 0.38" Maximum Flow Rate (Lpm): 8 Maximum Pressure (Psi): 60
Catalog No. NC0087851 Supplier PARKER HANNIFIN CORP Supplier No. 080694PSX5
The Scilog� Scipres� Combines Pressure Sensing Capabilities And The Convenience Of Disposability With Easy Setup. Each Sensor Is Preprogrammed With A Unique Id For Easy Traceability And Data Documentation When Combined With The Scilog� Scidoc Software. Factory Calibration Data Is Also Stored On Each Sensor'S Chip For Out-Of-Box, Plug And Play Use. Connection Type: 3/8" Barb Tube Size Range: Id 0.31" To 0.38" Maximum Flow Rate (Lpm): 8 Maximum Pressure (Psi): 60