Learn More
Med Vet International Universal Bandage Scissors, 7", German Grade, Von Klaus
Catalog No. 503046797
Encompass_Preferred
7" Universal Bandage Scissors is a German grade surgical instrument that has black handles. The tip has a flat blunt nodule to help slide the scisssors between bandages and skin. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-304-6797 Supplier Med Vet International Supplier No. VK1404718
7" Universal Bandage Scissors is a German grade surgical instrument that has black handles. The tip has a flat blunt nodule to help slide the scisssors between bandages and skin. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.