Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Scissor Blades Sharp
SDP

Catalog No. 21010359
Change view
Click to view available options
Material:
Stainless Steel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
21-010-359 Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-010-359 Supplier StatLab Supplier No. SIS1301P
This item is not returnable. View return policy

Sharp Scissor blades for various applications.

  • Sharp scissor blades
  • 5 Pk.

Specifications

Product Type Scissor Blades
Blade Material Metal
Blade Style Straight
Color Code Gray
Color-coded No
Grade General Precision
Handle Material Metal
Length (English) 2 in.
Length (Metric) 55 mm
Length (Metric) Blade Unknown
Material Stainless Steel
Size Standard
No. per Pack 5 Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.