Learn More
Med Vet International Miltex Darling Eccentric Bandage Scissors, Heavy Duty, Serrated, German, 6.25in, Pm-4126
Catalog No. 502829299
Encompass_Preferred
Durable, lightweight, dependable, and designed for cutting thick dressing material like bandages or gauze. 6.25in in length. Serrated finish on blades for reduced chance of slipping. Made with high-quality, medical-grade German steel.
Catalog No. 50-282-9299 Supplier Med Vet International Supplier No. MILPM4126
Durable, lightweight, dependable, and designed for cutting thick dressing material like bandages or gauze. 6.25in in length. Serrated finish on blades for reduced chance of slipping. Made with high-quality, medical-grade German steel.