Learn More
Med Vet International Miltex Mayo Dissecting Scissors, Straight, Rounded Blades, German, 5.75in, 5-136
Catalog No. 503045680
Encompass_Preferred
Thicker and more heavy duty than most, Mayo scissors are generally used near the surface of a surgical wound to cut fascia and muscle layers. It is standard for Mayo scissors to have standard or extra-long shafts. 5.75in in length. Straight rounded blades. Made with high-quality, medical grade German metal.
Catalog No. 50-304-5680 Supplier Med Vet International Supplier No. MIL5136
Thicker and more heavy duty than most, Mayo scissors are generally used near the surface of a surgical wound to cut fascia and muscle layers. It is standard for Mayo scissors to have standard or extra-long shafts. 5.75in in length. Straight rounded blades. Made with high-quality, medical grade German metal.