Learn More
Med Vet International 5.5-in Mayo Dissecting Scissor, Straight, Von Klaüs German Surgical Steel
Catalog No. 503046422
Encompass_Preferred
Mayo dissecting scissors are typically used for cutting surface tissue or sutures, The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-304-6422 Supplier Med Vet International Supplier No. VK1035014
Mayo dissecting scissors are typically used for cutting surface tissue or sutures, The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.