Learn More
Med Vet International SCISSORS, EAR CROPPING, 6.5", B/B, GERMAN
Catalog No. 503046421
Encompass_Preferred
Von Klaus curved Iris Scissors is a German Grade surgical instrument and feature have a sharp/sharp tip. Iris Scissors were originally designed for ophthalmic procedures but have become a very multipurpose tool. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-304-6421 Supplier Med Vet International Supplier No. VK1020166
Von Klaus curved Iris Scissors is a German Grade surgical instrument and feature have a sharp/sharp tip. Iris Scissors were originally designed for ophthalmic procedures but have become a very multipurpose tool. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.