Learn More
Fine Science Tools Fine Scissors, Large Loops, Stainless Steel, Curved, 10 cm
SDP
Catalog No. NC9708784
Encompass
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 25mm/ Alloy / Material: Stainless Steel/ Length: 10cm/ Serrated: No/ Feature: Large Loops
Catalog No. NC9708784 Supplier Fine Science Tools Supplier No. 1404110
Tip Shape: Curved/ Tips: Sharp-Sharp/ Cutting Edge: 25mm/ Alloy / Material: Stainless Steel/ Length: 10cm/ Serrated: No/ Feature: Large Loops