Learn More
Grainger Southworth Scissor Lift Table, Manual, 300 lb Capacity, 17 3/4 in. Max Height, Aluminum, 1 ea.
Catalog No. 502786478
Encompass_Preferred
This manual scissor lift table is designed for the efficient lifting and vertical positioning of equipment and materials in various industrial environments. Featuring a 300 lb load capacity and manual hand lever operation, this unit provides a reliable solution for applications where power sources are unavailable. Its stationary design and durable aluminum construction make it suitable for consistent workplace use.
  • Maximum load capacity of 300 lb
  • Maximum lifting height of 17 3/4 in
  • Minimum lifting height of 3 1/2 in
  • Manual operation via hand lever
  • Fixed aluminum platform with included ABS cover
  • Stationary, single-scissor design
  • Platform dimensions of 23 3/16 in L by 22 3/16 in W
Catalog No. 50-278-6478 Supplier Grainger Supplier No. 19A867
This manual scissor lift table is designed for the efficient lifting and vertical positioning of equipment and materials in various industrial environments. Featuring a 300 lb load capacity and manual hand lever operation, this unit provides a reliable solution for applications where power sources are unavailable. Its stationary design and durable aluminum construction make it suitable for consistent workplace use.
  • Maximum load capacity of 300 lb
  • Maximum lifting height of 17 3/4 in
  • Minimum lifting height of 3 1/2 in
  • Manual operation via hand lever
  • Fixed aluminum platform with included ABS cover
  • Stationary, single-scissor design
  • Platform dimensions of 23 3/16 in L by 22 3/16 in W