Learn More
Med Vet International Miltex Locklin Operating Scissors, Curved Shank, Angled On Side 25 Degrees, One Serrated Blade, German, 6.25in, 5D-328
Catalog No. 502832475
Encompass_Preferred
Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. Used to cut tissue, as well as other things such as bandages and gauze. Serrated blade helps prevent tissue slippage. Curved shank. Angled on side 25 degree. 6.25in total length. These scissors are used for general, multipurpose cutting and dissecting. Made with high-quality, medical grade German steel.
Catalog No. 50-283-2475 Supplier Med Vet International Supplier No. MIL5D329
Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. Used to cut tissue, as well as other things such as bandages and gauze. Serrated blade helps prevent tissue slippage. Curved shank. Angled on side 25 degree. 6.25in total length. These scissors are used for general, multipurpose cutting and dissecting. Made with high-quality, medical grade German steel.