Learn More
Med Vet International Miltex Mayo Scissors, Tungsten Carbide, Curved, Blunt/Blunt, German, 6.75in, 5-142Tc
Catalog No. 502831235
Encompass_Preferred
Thicker and more heavy duty than most, Mayo scissors are generally used near the surface of a surgical wound to cut fascia and muscle layers. It is standard for Mayo scissors to have standard or extra-long shafts. 6.75in in length. Curved blades with blunt/blunt tips. Tungsten carbide coated to ensure prolonged durability. Made with high-quality, medical grade German metal.
Catalog No. 50-283-1235 Supplier Med Vet International Supplier No. MIL5142TC
Thicker and more heavy duty than most, Mayo scissors are generally used near the surface of a surgical wound to cut fascia and muscle layers. It is standard for Mayo scissors to have standard or extra-long shafts. 6.75in in length. Curved blades with blunt/blunt tips. Tungsten carbide coated to ensure prolonged durability. Made with high-quality, medical grade German metal.