Learn More
Med Vet International Miltex Meisterhand Metzenbaum Scissors, Delicate, Curved, Blunt, German, 7in, Mh5-288
Catalog No. 502832256
Encompass_Preferred
Used for cutting delicate tissue and blunt dissection. Great for surgery and suture material. Equipped with extra-long handles and short-blades for deep cuts into the body. Kept in most operating rooms, surgical bays, and other medical settings. 7in in length. Curved blades with blunt tips making them ideal for biopsy and dissection. Made with medical grade German steel.
Catalog No. 50-283-2256 Supplier Med Vet International Supplier No. MILMH5288
Used for cutting delicate tissue and blunt dissection. Great for surgery and suture material. Equipped with extra-long handles and short-blades for deep cuts into the body. Kept in most operating rooms, surgical bays, and other medical settings. 7in in length. Curved blades with blunt tips making them ideal for biopsy and dissection. Made with medical grade German steel.