Learn More
Med Vet International Miltex Meisterhand Metzenbaum-Lahey, Straight, Blunt Points, German, 5.75in, Mh5-179
Catalog No. 502831907
Encompass_Preferred
Used for cutting delicate tissue and blunt dissection. Great for surgery and suture material. Equipped with extra-long handles and short-blades for deep cuts into the body. Kept in most operating rooms, surgical bays, and other medical settings. MeisterHand scissors ensure superior quality. Straight blades with blunt points. Standard pattern. 5.75in total length. Made with high-quality German steel.
Catalog No. 50-283-1907 Supplier Med Vet International Supplier No. MILMH5179
Used for cutting delicate tissue and blunt dissection. Great for surgery and suture material. Equipped with extra-long handles and short-blades for deep cuts into the body. Kept in most operating rooms, surgical bays, and other medical settings. MeisterHand scissors ensure superior quality. Straight blades with blunt points. Standard pattern. 5.75in total length. Made with high-quality German steel.