Learn More
Med Vet International Miltex Meisterhand Metzenbaum Scissors, Curved, Blunt Points, German, 7in, Mh5-182
Catalog No. 502831568
Encompass_Preferred
Used for cutting delicate tissue and blunt dissection. Great for surgery and suture material. Equipped with extra-long handles and short-blades for deep cuts into the body. Kept in most operating rooms, surgical bays, and other medical settings. MeisterHand scissors guarantee superior craftsmanship. 7in total length. Made with high-quality, medical grade German steel.
Catalog No. 50-283-1568 Supplier Med Vet International Supplier No. MILMH5182
Used for cutting delicate tissue and blunt dissection. Great for surgery and suture material. Equipped with extra-long handles and short-blades for deep cuts into the body. Kept in most operating rooms, surgical bays, and other medical settings. MeisterHand scissors guarantee superior craftsmanship. 7in total length. Made with high-quality, medical grade German steel.