Learn More
Med Vet International Miltex Metzenbaum-Lahey Scissors, Tungsten Carbide, Standard Pattern, Straight, Blunt, German, 5.75in, 5-179Tc
Catalog No. 502831824
Encompass_Preferred
Used for cutting delicate tissue and blunt dissection. Great for surgery and suture material. Gold-plated handles and tungsten carbide inlays bonded to the scissor blades insure increased durability, sharper blades, and extended service life. Equipped with extra-long handles and short-blades for deep cuts into the body. Kept in most operating rooms, surgical bays, and other medical settings. Straight blade with blunt points. 5.75in total length. Made with high-quality, medical grade German steel.
Catalog No. 50-283-1824 Supplier Med Vet International Supplier No. MIL5179TC
Used for cutting delicate tissue and blunt dissection. Great for surgery and suture material. Gold-plated handles and tungsten carbide inlays bonded to the scissor blades insure increased durability, sharper blades, and extended service life. Equipped with extra-long handles and short-blades for deep cuts into the body. Kept in most operating rooms, surgical bays, and other medical settings. Straight blade with blunt points. 5.75in total length. Made with high-quality, medical grade German steel.