Learn More
Med Vet International Operating Scissors, 5", Curved, Blunt/Blunt Tip, German Grade, Von Klaus
Catalog No. 503046047
Encompass_Preferred
5" German Grade Curved Operating Scissors are a versatile surgical isntrument. These Surgical Scissors are curved and have a blunt/blunt tip and are used generally for cutting sutures. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.”
Catalog No. 50-304-6047 Supplier Med Vet International Supplier No. VK1030313
5" German Grade Curved Operating Scissors are a versatile surgical isntrument. These Surgical Scissors are curved and have a blunt/blunt tip and are used generally for cutting sutures. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.”