Learn More
Med Vet International Operating Scissors, 6.5", Straight, Sharp/Blunt Tip, German Grade, Von Klaus
Catalog No. 503045604
Encompass_Preferred
6.5" German Grade straight operating scissors are a versatile surgical isntrument. These Surgical Scissors are straight and have a sharp/blunt tip and are used generally for cutting and dissecting. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-304-5604 Supplier Med Vet International Supplier No. VK1030616
6.5" German Grade straight operating scissors are a versatile surgical isntrument. These Surgical Scissors are straight and have a sharp/blunt tip and are used generally for cutting and dissecting. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.