Learn More
Med Vet International Miltex Operating Scissors, Delicate Pattern, Lightweight, Curved, Blunt/Blunt, German, 5.5in, 5-90
Catalog No. 502831274
Encompass_Preferred
Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. Used to cut tissue, as well as other things such as bandages and gauze. These scissors are used for general, multipurpose cutting and dissecting. 5.5in in length. Light in weight to prevent prolonged fatigue. Curved blades with blunt/blunt tips. Made with high-quality, medical grade German steel.
Catalog No. 50-283-1274 Supplier Med Vet International Supplier No. MIL590
Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. Used to cut tissue, as well as other things such as bandages and gauze. These scissors are used for general, multipurpose cutting and dissecting. 5.5in in length. Light in weight to prevent prolonged fatigue. Curved blades with blunt/blunt tips. Made with high-quality, medical grade German steel.