Learn More
Med Vet International Miltex Operating Scissors, Standard Pattern, Curved, 5.5in, Mh5-46
Catalog No. 502831363
Encompass_Preferred
Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. Used to cut tissue, as well as other things such as bandages and gauze. These scissors are used for general, multipurpose cutting and dissecting. MeisterHand scissors guarantee excellent craftsmanship. Curved blades. 5.5in total length. Made with high-quality, medical grade German steel.
Catalog No. 50-283-1363 Supplier Med Vet International Supplier No. MILMH546
Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. Used to cut tissue, as well as other things such as bandages and gauze. These scissors are used for general, multipurpose cutting and dissecting. MeisterHand scissors guarantee excellent craftsmanship. Curved blades. 5.5in total length. Made with high-quality, medical grade German steel.