Learn More
Med Vet International Miltex Post Mortem Dissecting Scissors, Straight, Sharp/Sharp Points, 4.5in, 34-80
Catalog No. 502831143
Encompass_Preferred
Used for performing autopsy procedures. Larger blades make it easier to cut through tissues and bones. Straight blades with sharp/sharp tips. 4.5in total length. Made with high-quality, medical-grade German steel.
Catalog No. 50-283-1143 Supplier Med Vet International Supplier No. MIL3480
Used for performing autopsy procedures. Larger blades make it easier to cut through tissues and bones. Straight blades with sharp/sharp tips. 4.5in total length. Made with high-quality, medical-grade German steel.