Learn More
Med Vet International Miltex Plastic & ReconstructIVe Dissecting & Tissue Scissors, Curved, Supercut, 4.5in, 21-Sc-745
Catalog No. 502830084
Encompass_Preferred
Designed specifically for cosmetic and plastic surgery. They are very delicate and lightweight making them ideal for dissecting, cutting, holding and suturing tissue during restorative surgery. Black rubber grips on the rings provide a secure grip. Curved SuperCut blades. 4.5in total length. Made with high-quality, medical-grade German steel.
Catalog No. 50-283-0084 Supplier Med Vet International Supplier No. MIL21SC745
Designed specifically for cosmetic and plastic surgery. They are very delicate and lightweight making them ideal for dissecting, cutting, holding and suturing tissue during restorative surgery. Black rubber grips on the rings provide a secure grip. Curved SuperCut blades. 4.5in total length. Made with high-quality, medical-grade German steel.