Learn More
Med Vet International Miltex Rees Rhytidectomy Scissors, Curved, Serrated, German, 6.25in, Pm-6575
Catalog No. 502830767
Encompass_Preferred
Rhytidectomy scissors offer extended shaft lengths for greater surgical visualization. Featured are specially-designed razor sharp upper blade edges to cut cleanly and quickly though dense or delicate tissue. Curved blades with a serrated finish to prevent slips. 6.25in total length. Made with high-quality, medical grade German steel.
Catalog No. 50-283-0767 Supplier Med Vet International Supplier No. MILPM6575
Rhytidectomy scissors offer extended shaft lengths for greater surgical visualization. Featured are specially-designed razor sharp upper blade edges to cut cleanly and quickly though dense or delicate tissue. Curved blades with a serrated finish to prevent slips. 6.25in total length. Made with high-quality, medical grade German steel.