Learn More
Med Vet International Sims Uterine Scissors, Sharp / Blunt, Straight, 8", Ea
Catalog No. 502831927
Encompass_Preferred
Sims Uterine Scissors Overall Length (203cm)Straight or Curved Sharp or Blunt Specifications INSTRUMENT FAMILY Sims INSTRUMENT GRADE German Grade Stainless Steel LATEX FREE Yes LENGTH IN CM20.3 cm SPECIALTY USE Obstetrics/Gynecology TYPE OF INSTRUMENT Uterine ScissorsUNSPSC42291614
Catalog No. 50-283-1927 Supplier Med Vet International Supplier No. MDS0851220
Sims Uterine Scissors Overall Length (203cm)Straight or Curved Sharp or Blunt Specifications INSTRUMENT FAMILY Sims INSTRUMENT GRADE German Grade Stainless Steel LATEX FREE Yes LENGTH IN CM20.3 cm SPECIALTY USE Obstetrics/Gynecology TYPE OF INSTRUMENT Uterine ScissorsUNSPSC42291614