Learn More
Med Vet International Miltex Spencer Stitch Scissors, 3.5in, 12 Pack, Eg9-100
Catalog No. 502829780
Encompass_Preferred
Primarily used for suture removal. The hook tip helps the blades slide under sutures for an easy and safe process. Professionally sterilized for practicality, ease, and safety. 3.5in total length. Made with high-quality, medical-grade German steel.
Catalog No. 50-282-9780 Supplier Med Vet International Supplier No. MILEG9100
Primarily used for suture removal. The hook tip helps the blades slide under sutures for an easy and safe process. Professionally sterilized for practicality, ease, and safety. 3.5in total length. Made with high-quality, medical-grade German steel.