Learn More
Med Vet International Miltex Spencer Stitch Scissors, Curved On Flat, German, 3.5in, Pm-6751
Catalog No. 502830057
Encompass_Preferred
Primarily used for suture removal. The hook tip helps the blades slide under sutures for an easy and safe process, and the flat orientation allows additional maneuverability. Professionally sterilized for practicality, ease, and safety. 3.5in total length. Made with high-quality, medical-grade German steel.
Catalog No. 50-283-0057 Supplier Med Vet International Supplier No. MILPM6751
Primarily used for suture removal. The hook tip helps the blades slide under sutures for an easy and safe process, and the flat orientation allows additional maneuverability. Professionally sterilized for practicality, ease, and safety. 3.5in total length. Made with high-quality, medical-grade German steel.