Learn More
Med Vet International Miltex Utility Scissors, Curved Blunt Tips, Ribbon Type, German, 3.87in, 5-400
Catalog No. 502832258
Encompass_Preferred
Scissors designed for use in medical, surgical, or laboratory settings. Made to be used in a number of applications, such as surgical, education, veterinary, and much more. Westcott type scissors are delicate for finer incisions. Curved blades with blunt tips. 3.87in total length. Put together with high-quality, medical grade German steel.
Catalog No. 50-283-2258 Supplier Med Vet International Supplier No. MIL5400
Scissors designed for use in medical, surgical, or laboratory settings. Made to be used in a number of applications, such as surgical, education, veterinary, and much more. Westcott type scissors are delicate for finer incisions. Curved blades with blunt tips. 3.87in total length. Put together with high-quality, medical grade German steel.