Learn More
Med Vet International Miltex Vannas Capsulotomy Scissors, Curved, Sharp Tips, Extra Delicate, German, 3.25in, Mh18-1622
Supplier:  Med Vet International MILMH181622
Encompass_Preferred
Dynarex Safe-Touch Blue Nitrile Exam Gloves are premium-quality powder-free nitrile gloves crafted for optimal protection and comfort. These ambidextrous Gloves are designed with textured fingertips for improved grip. The Nitrile Exam Gloves feature a thickness range of 3-4 mil at the palm and 4-6 mil at fingertips. The Exam Gloves feature a polymer lining and beaded cuff to help ensure easy donning and help prevent tears. These Gloves have a secure fit around the wrist and h and -conforming flexibility to help prevent physical fatigue. Dynarex Safe-Touch Blue Nitrile Exam Gloves are high-quality polymer-lined gloves designed for enhanced protection and performance. The Nitrile Exam Gloves have a thickness range of 3-4 mil at the palm and 4-6 mil at fingertips to provide optimal tactile sensitivity and puncture resistance. These ambidextrous powder-free Gloves feature textured fingertips for improved grip and beaded cuffs to allow for easy donning.  
Catalog No. 50-283-1499