Learn More
Med Vet International Miltex Vannas Capsulotomy Scissors, Angled On Flat, Sharp Tips, German, 3.25in, 18-1624
Catalog No. 502829146
Encompass_Preferred
Operating room grade ultra-fine sharp-tip scissors. Angled blades on flat with sharp tips. 3.25in total length. Ideal for ophthalmic, corneal, and optical applications. Made with high-quality German steel.
Catalog No. 50-282-9146 Supplier Med Vet International Supplier No. MIL181624
Operating room grade ultra-fine sharp-tip scissors. Angled blades on flat with sharp tips. 3.25in total length. Ideal for ophthalmic, corneal, and optical applications. Made with high-quality German steel.