Learn More
Roboz Surgical SCISSOR VET HVY PATTERN
SDP
Catalog No. NC9506363
Encompass
Veterinary Scissors; Heavy Pattern; Straight, Sharp-Blunt Serrated Blades; Blade Length 57mm 6-1/2 in. Overall Length.
Catalog No. NC9506363 Supplier Roboz Surgical Supplier No. RS6940
Veterinary Scissors; Heavy Pattern; Straight, Sharp-Blunt Serrated Blades; Blade Length 57mm 6-1/2 in. Overall Length.