Learn More
Andwin Scientific SCISSOR W/REPL BLADES 4794
SDP
Catalog No. NC9821139
Encompass_Preferred
The Tissue-Tek Accu-Edge replaceable blade scissors are ideal tool for performing anatomical sectioning of tissues. The blades can be easily inserted and removed, which removes the time, expense, and incovenience of sharpening. This product is autoclavable and is sold individually.
Catalog No. NC9821139 Supplier Andwin Scientific Supplier No. 4794
The Tissue-Tek Accu-Edge replaceable blade scissors are ideal tool for performing anatomical sectioning of tissues. The blades can be easily inserted and removed, which removes the time, expense, and incovenience of sharpening. This product is autoclavable and is sold individually.