Learn More
Med Vet International Iris Scissors, 3.5in, Straight, Sharp/Sharp Tip, German Grade, Von Klaus
Catalog No. 502092702
Encompass_Preferred
3.5' German Grade Straight Iris Scissors are straight and have a sharp/sharp tip. Iris scissors were designed for ophthalmic procedures but have now become a multipurpose surgical instrument. Iris scissors are ideally used for dissection of fine tissue or fine suture removal. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-2702 Supplier Med Vet International Supplier No. VK1022009
3.5' German Grade Straight Iris Scissors are straight and have a sharp/sharp tip. Iris scissors were designed for ophthalmic procedures but have now become a multipurpose surgical instrument. Iris scissors are ideally used for dissection of fine tissue or fine suture removal. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.