Learn More
Med Vet International 5.5-in Mayo Dissecting Scissors, Curved, Von Klaüs German Surgical Steel
Catalog No. 502092704
Encompass_Preferred
Mayo dissecting scissors are made of high quality German surgical steel. These dissecting scissors are curved and are commonly used for cutting or dissecting dense tissue. Dissecting scissors are a very versatile instrument that can be used in multiple procedures. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose
Catalog No. 50-209-2704 Supplier Med Vet International Supplier No. VK1035114
Mayo dissecting scissors are made of high quality German surgical steel. These dissecting scissors are curved and are commonly used for cutting or dissecting dense tissue. Dissecting scissors are a very versatile instrument that can be used in multiple procedures. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose