Learn More
Med Vet International Metzenbaum Dissecting Scissors, 5.75in, Standard Pattern, Straight, German Grade, Von Klaus
Supplier:  Med Vet International VK1043214
Encompass_Preferred
5 3/4in. German grade Metzenbaum dissecting scissors are commonly used for dissecting or cutting tissue. They are straight and have a standard pattern. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-2711