Learn More
Med Vet International Operating Scissors, 4in, Curved, Sharp/Sharp Tip, German Grade, Von Klaus
Catalog No. 502092720
Encompass_Preferred
4' German Grade curved operating scissors are a versatile surgical isntrument. These Surgical Scissors are curved and have a sharp/sharp tip and are used generally for cutting tougher tissue. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-2720 Supplier Med Vet International Supplier No. VK1030511
4' German Grade curved operating scissors are a versatile surgical isntrument. These Surgical Scissors are curved and have a sharp/sharp tip and are used generally for cutting tougher tissue. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.