Learn More
Fine Science Tools Round Handled Vannas Spring Scissors, Stainless Steel, 12.5 cm
SDP
Catalog No. NC0801404
Encompass
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 6mm/ Tip Diameter: 0.075mm/ Alloy / Material: Stainless Steel/ Length: 12.5cm/ Serrated: No
Catalog No. NC0801404 Supplier Fine Science Tools Supplier No. 1540012
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 6mm/ Tip Diameter: 0.075mm/ Alloy / Material: Stainless Steel/ Length: 12.5cm/ Serrated: No