Learn More
Fine Science Tools Vannas-Tübingen Spring Scissors, Stainless Steel, 5 mm Cutting Edge, 8.5 cm
SDP
Catalog No. NC9308617
Encompass
Tip Shape: Angled to Side/ Tips: Sharp/ Cutting Edge: 5mm/ Tip Diameter: 0.075mm/ Alloy / Material: Stainless Steel/ Length: 8.5cm/ Serrated: No
Catalog No. NC9308617 Supplier Fine Science Tools Supplier No. 1500808
Tip Shape: Angled to Side/ Tips: Sharp/ Cutting Edge: 5mm/ Tip Diameter: 0.075mm/ Alloy / Material: Stainless Steel/ Length: 8.5cm/ Serrated: No